Ethyl indole-2-carboxylate, 98+%, Thermo Scientific Chemicals
Ethyl indole-2-carboxylate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl indole-2-carboxylate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L00797.14
also known as L00797-14
Price (USD)/ Each
161.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
161.00
Add to cart
Ethyl indole-2-carboxylate, 98+%, Thermo Scientific Chemicals
Catalog numberL00797.14
Price (USD)/ Each
161.00
-
Add to cart
Chemical Identifiers
CAS3770-50-1
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Melting Point (clear melt)120.0-126.0?C
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only