Thermo Scientific Chemicals

1-Benzylimidazole, 98+%, Thermo Scientific Chemicals

Catalog number: L00802.14
25 g, Each
Thermo Scientific Chemicals

1-Benzylimidazole, 98+%, Thermo Scientific Chemicals

Catalog number: L00802.14
25 g, Each
Quantity
Catalog number: L00802.14
also known as L00802-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4238-71-5
IUPAC Name
1-benzyl-1H-imidazole
Molecular Formula
C10H10N2
InChI Key
KKKDZZRICRFGSD-UHFFFAOYSA-N
SMILES
C(N1C=CN=C1)C1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥98.0%

Description

1-Benzylimidazole acts as an inducer of various cytochrome P-450 isozymes and inhibitor of thromboxane A2 synthase. 1-Benzylimidazole has been used to prepare cyclodextrin-ionic liquid polymer (βCD-BIMOTs-TDI)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzylimidazole acts as an inducer of various cytochrome P-450 isozymes and inhibitor of thromboxane A2 synthase. 1-Benzylimidazole has been used to prepare cyclodextrin-ionic liquid polymer (βCD-BIMOTs-TDI)

Solubility
Soluble in methanol, acetone, DMSO, ethanol, dilute acids (freely soluble), and buffers of pH>6.0 (freely soluble). Insoluble in water.

Notes
Store in a cool, dry place. Keep container closed when not in use. Incompatible with Oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text