alpha-Methylcinnamic acid, 99%, Thermo Scientific Chemicals
alpha-Methylcinnamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

alpha-Methylcinnamic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L00806.06
also known as L00806-06
Price (USD)/ Each
36.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
36.60
Add to cart
alpha-Methylcinnamic acid, 99%, Thermo Scientific Chemicals
Catalog numberL00806.06
Price (USD)/ Each
36.60
-
Add to cart
Chemical Identifiers
CAS1199-77-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.5%
Melting Point (clear melt)77.5-83.5?C
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only