Additional water and monomethyl hemiacetal also present
Comment
May form oligomers and cyclic trimer
Form
Fused solid, clear as melt
Assay (unspecified)
>88.0% (as gem-diol/aldehyde monohydrate)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.