2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.
Solubility
Slightly Soluble (1.2 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
DT Gibson.; JR Koch.; CL Schuld.; RE Kallio. Oxidative degradation of aromatic hydrocarbons by microorganisms. II. Metabolism of halogenated aromatic hydrocarbons. Biochemistry. 19687 (11) , 3795-3802.
FA Pasha.; HK Srivastava.; PP Singh. Comparative QSAR study of phenol derivatives with the help of density functional theory. Bioorganic & medicinal chemistry. 200513 (24) , 6823-6829.