Thermo Scientific Chemicals

2-Chloro-5-methylphenol, 99%, Thermo Scientific Chemicals

Catalog number: L00816.22
100 g, Each
Thermo Scientific Chemicals

2-Chloro-5-methylphenol, 99%, Thermo Scientific Chemicals

Catalog number: L00816.22
100 g, Each
Quantity
Catalog number: L00816.22
also known as L00816-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
615-74-7
IUPAC Name
2-chloro-5-methylphenol
Molecular Formula
C7H7ClO
InChI Key
SMFHPCZZAAMJJO-UHFFFAOYSA-N
SMILES
CC1=CC=C(Cl)C(O)=C1
Appearance (Color)
White to cream to yellow to pale brown
Form
Crystals or powder or crystalline powder or fused solid or chunks
Assay (GC)
≥98.5%
Melting Point (clear melt)
43.0-49.0?C

Description

2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.

Solubility
Slightly Soluble (1.2 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text