2-Benzimidazoleacetonitrile, is used as a pharmaceutical intermediate. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Benzimidazoleacetonitrile, is used as a pharmaceutical intermediate. It is also used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
General References:
- VD Dyachenko.; OS Bityukova. Regioselectivity of heterocyclization of 2-substituted 3, 3-bis (methylsulfanyl) acrylo-nitriles with 2-benzimidazoleacetonitrile. Chemistry of Heterocyclic Compounds. 201046 (8) , 938-940.
- N Finch.; CW Gemenden. Heterocyclic synthesis with 2-benzimidazoleacetic acid derivatives. The Journal of Organic Chemistry. 197035(9) , 3114-3119.