Thermo Scientific Chemicals

Phenyl chlorothionoformate, 98+%, Thermo Scientific Chemicals

Catalog number: L00838.06
5 g, Each
Thermo Scientific Chemicals

Phenyl chlorothionoformate, 98+%, Thermo Scientific Chemicals

Catalog number: L00838.06
5 g, Each
Quantity
Catalog number: L00838.06
also known as L00838-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1005-56-7
IUPAC Name
phenyl chloromethanethioate
Molecular Formula
C7H5ClOS
InChI Key
KOSYAAIZOGNATQ-UHFFFAOYSA-N
SMILES
ClC(=S)OC1=CC=CC=C1
Assay (GC)
≥98.0%
Appearance (Color)
Clear yellow
Form
Liquid
Refractive Index
1.5800-1.5830 @ 20?C

Description

Phenyl chlorothionoformate has been used in stereo directed synthesis of optically active, (-)-mintlactone, synthesis of peptide α-thioesters having a variety of C-terminal amino acids, synthesis of scyllo-inositol derivatives, thionocarbonylation of unprotected thymine nucleosides, preparation of phenoxythiocarbonyl esters of protected ribonucleosides. It is also used as a reagent in the thionocarbonylation of unprotected thymine nucleosides. It forms phenoxythiocarbonyl esters of protected ribonucleosides which can be reduced by tributyltin hydride to deoxyribonucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl chlorothionoformate has been used in stereo directed synthesis of optically active, (-)-mintlactone, synthesis of peptide α-thioesters having a variety of C-terminal amino acids, synthesis of scyllo-inositol derivatives, thionocarbonylation of unprotected thymine nucleosides, preparation of phenoxythiocarbonyl esters of protected ribonucleosides. It is also used as a reagent in the thionocarbonylation of unprotected thymine nucleosides. It forms phenoxythiocarbonyl esters of protected ribonucleosides which can be reduced by tributyltin hydride to deoxyribonucleosides.

Solubility
Soluble in water.

Notes
Moisture, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text