5-Hydroxyindole, 98+%, Thermo Scientific Chemicals
5-Hydroxyindole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Hydroxyindole, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L00858.06
also known as L00858-06
Price (USD)/ Each
217.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
217.00
Add to cart
5-Hydroxyindole, 98+%, Thermo Scientific Chemicals
Catalog numberL00858.06
Price (USD)/ Each
217.00
-
Add to cart
Chemical Identifiers
CAS1953-54-4
IUPAC Name1H-indol-5-ol
Molecular FormulaC8H7NO
InChI KeyLMIQERWZRIFWNZ-UHFFFAOYSA-N
SMILESOC1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.0%
Appearance (Color)White to cream to brown to pale gray to gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only