Phenylpropiolaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
Phenylpropiolaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenylpropiolaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L00872.14
also known as L00872-14
Price (USD)/ Each
96.80
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
96.80
Add to cart
Phenylpropiolaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
Catalog numberL00872.14
Price (USD)/ Each
96.80
-
Add to cart
Chemical Identifiers
CAS6142-95-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5150-1.5190 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only