Thermo Scientific Chemicals

Methylamine, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: L00894.AU
1000 mL, Each
Thermo Scientific Chemicals

Methylamine, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: L00894.AU
1000 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L00894.AU
also known as L00894-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74-89-5
IUPAC Name
methanamine
Molecular Formula
CH5N
InChI Key
BAVYZALUXZFZLV-UHFFFAOYSA-N
SMILES
CN
Appearance (Color)
Clear colorless
Form
Liquid
Assay (Non-aqueous acid-base Titration)
37-43% w/w

Description

Methylamine, 40% w/w aq. solution is used as a mobile phase in the perchlorate analysis of milk and yogurt samples by LC-MS/MS. It is also used in the synthesis of 9-fluorenylmethyl N-methylcarbamate and tert-butyl N-methylcarbamate, 7-methylamino-2-(2-quinolyl)tropone, 2,5-meso-pyrrolidines and 2-(4-chlorophenyl)-N-methylacetamide. This compound also serves as a building block for the synthesis of many other commercially available compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text