Thermo Scientific Chemicals

Ethoxycarbonyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L00911.06
5 g, Each
Thermo Scientific Chemicals

Ethoxycarbonyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L00911.06
5 g, Each
Quantity
Catalog number: L00911.06
also known as L00911-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16182-04-0
IUPAC Name
ethyl N-carbothioylcarbamate
Molecular Formula
C4H5NO2S
InChI Key
BDTDECDAHYOJRO-UHFFFAOYSA-N
SMILES
CCOC(=O)N=C=S
Appearance (Color)
Clear yellow to orange-brown
Assay (GC)
>96.0%
Refractive Index
1.4975-1.5025 @ 20?C
Form
Liquid

Description

Ethoxycarbonyl isothiocyanate, is used as a building block in the synthesis of various heterocycles. It is also used for the synthesis of 1,2,4-oxadiazolidine-3,5-dione. It is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethoxycarbonyl isothiocyanate, is used as a building block in the synthesis of various heterocycles. It is also used for the synthesis of 1,2,4-oxadiazolidine-3,5-dione. It is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform and ether.

Notes
Moisture, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text