Thermo Scientific Chemicals

Phenyl carbamate, 98+%, Thermo Scientific Chemicals

Catalog number: L00944.14
25 g, Each
Thermo Scientific Chemicals

Phenyl carbamate, 98+%, Thermo Scientific Chemicals

Catalog number: L00944.14
25 g, Each
Quantity
Catalog number: L00944.14
also known as L00944-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
622-46-8
IUPAC Name
phenyl carbamate
Molecular Formula
C7H7NO2
InChI Key
BSCCSDNZEIHXOK-UHFFFAOYSA-N
SMILES
NC(=O)OC1=CC=CC=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused solid
Assay (HPLC)
≥98.0%
Identification (FTIR)
Conforms

Description

Phenyl carbamate, is used as various phenyl carbamate esters derived from amino acids, a dipeptide and amino acid esters and amides were prepared and assessed as potential prodrugs with the aim of protecting phenolic drugs against first-pass metabolism following per oral administration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl carbamate, is used as various phenyl carbamate esters derived from amino acids, a dipeptide and amino acid esters and amides were prepared and assessed as potential prodrugs with the aim of protecting phenolic drugs against first-pass metabolism following per oral administration.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text