Thermo Scientific Chemicals

Triphenylene, 98%, Thermo Scientific Chemicals

Catalog number: L00955.03
1 g, Each
Thermo Scientific Chemicals

Triphenylene, 98%, Thermo Scientific Chemicals

Catalog number: L00955.03
1 g, Each
Quantity
Catalog number: L00955.03
also known as L00955-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
217-59-4
IUPAC Name
triphenylene
Molecular Formula
C18H12
InChI Key
SLGBZMMZGDRARJ-UHFFFAOYSA-N
SMILES
C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21
Assay (HPLC)
≥97.5%
Form
Crystals or powder or crystalline powder or granules
Appearance (Color)
White to cream to yellow to pale brown

Description

Triphenylene is used in optics and electronics. It is also used as a discotic mesogen in liquid crystalline materials. Further, it is a compound that fluoresces in the ultraviolet region. In addition to this, it is used in the preparation of triphenylene-2-carbaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylene is used in optics and electronics. It is also used as a discotic mesogen in liquid crystalline materials. Further, it is a compound that fluoresces in the ultraviolet region. In addition to this, it is used in the preparation of triphenylene-2-carbaldehyde.

Solubility
Soluble in ethanol, benzene, acetic acid, oils and chloroform.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text