Triphenylene is used in optics and electronics. It is also used as a discotic mesogen in liquid crystalline materials. Further, it is a compound that fluoresces in the ultraviolet region. In addition to this, it is used in the preparation of triphenylene-2-carbaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triphenylene is used in optics and electronics. It is also used as a discotic mesogen in liquid crystalline materials. Further, it is a compound that fluoresces in the ultraviolet region. In addition to this, it is used in the preparation of triphenylene-2-carbaldehyde.
Solubility
Soluble in ethanol, benzene, acetic acid, oils and chloroform.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Collins, K. D.; Honeker, R.; Vásquez-Céspedes, S.; Tang, D. T. D.; Glorius, F. C-H arylation of triphenylene, naphthalene and related arenes using Pd/C. Chem. Sci. 2015, 6, 1816-1824.
Reinhard, D.; Rominger, F.; Mastalerz, M. Synthesis of Triphenylene-Based Triptycenes via Suzuki-Miyaura Cross-Coupling and Subsequent Scholl Reaction. J. Org. Chem. 2015, 80 (18), 9342-9348.