Thermo Scientific Chemicals

Glutarimide, 98%, Thermo Scientific Chemicals

Catalog number: L00968.22
100 g, Each
Thermo Scientific Chemicals

Glutarimide, 98%, Thermo Scientific Chemicals

Catalog number: L00968.22
100 g, Each
Quantity
Catalog number: L00968.22
also known as L00968-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1121-89-7
IUPAC Name
piperidine-2,6-dione
Molecular Formula
C5H7NO2
InChI Key
KNCYXPMJDCCGSJ-UHFFFAOYSA-N
SMILES
O=C1CCCC(=O)N1
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
148-159?C
Assay (GC)
≥97.5%
Appearance (Color)
White
Form
Crystals or crystalline powder or flakes

Description

Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.

Solubility
Soluble in water, hot ethanol and boiling benzene. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text