3-Methylbiphenyl, 95%, Thermo Scientific Chemicals
3-Methylbiphenyl, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylbiphenyl, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L01006.06
also known as L01006-06
Price (USD)/ Each
181.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
181.00
Add to cart
3-Methylbiphenyl, 95%, Thermo Scientific Chemicals
Catalog numberL01006.06
Price (USD)/ Each
181.00
-
Add to cart
Chemical Identifiers
CAS643-93-6
IUPAC Name3-methyl-1,1'-biphenyl
Molecular FormulaC13H12
InChI KeyNPDIDUXTRAITDE-UHFFFAOYSA-N
SMILESCC1=CC=CC(=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid or viscous liquid
Refractive Index1.5990-1.6050 @ 20?C
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only