Thermo Scientific Chemicals

4-Hydroxy-3,5-diiodobenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: L01060.18
50 g, Each
Thermo Scientific Chemicals

4-Hydroxy-3,5-diiodobenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: L01060.18
50 g, Each
Quantity
Catalog number: L01060.18
also known as L01060-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1948-40-9
IUPAC Name
4-hydroxy-3,5-diiodobenzaldehyde
Molecular Formula
C7H4I2O2
InChI Key
WHLUEIMENHLCMY-UHFFFAOYSA-N
SMILES
OC1=C(I)C=C(C=O)C=C1I
Appearance (Color)
White to cream to orange or pale yellow or pale brown
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.0%
Melting Point (clear melt)
198-207?C

Description

4-Hydroxy-3,5-diiodobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,5-diiodobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water. Sparingly soluble in chloroform and methanol.

Notes
Air & Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text