3-Chlorobenzhydrazide, 98+%, Thermo Scientific Chemicals
3-Chlorobenzhydrazide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chlorobenzhydrazide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L01071.18
also known as L01071-18
Price (USD)/ Each
396.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
396.00
Add to cart
3-Chlorobenzhydrazide, 98+%, Thermo Scientific Chemicals
Catalog numberL01071.18
Price (USD)/ Each
396.00
-
Add to cart
Chemical Identifiers
CAS1673-47-8
IUPAC Name3-chlorobenzohydrazide
Molecular FormulaC7H7ClN2O
InChI KeyPHRDZSRVSVNQRN-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or fused solid or needles
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only