Thermo Scientific Chemicals

4-Oxo-TEMPO, free radical, 96%, Thermo Scientific Chemicals

Catalog number: L01084.03
1 g, Each
Thermo Scientific Chemicals

4-Oxo-TEMPO, free radical, 96%, Thermo Scientific Chemicals

Catalog number: L01084.03
1 g, Each
Quantity
Catalog number: L01084.03
also known as L01084-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2896-70-0
IUPAC Name
(2,2,6,6-tetramethyl-4-oxopiperidin-1-yl)oxidanyl
Molecular Formula
C9H16NO2
InChI Key
WSGDRFHJFJRSFY-UHFFFAOYSA-N
SMILES
CC1(C)CC(=O)CC(C)(C)N1[O]
Form
Crystals or powder or crystalline powder
Assay (GC)
≥95.0%
Appearance (Color)
Yellow to orange
Melting Point (clear melt)
33.0-42.0?C

Description

4-Oxo-Tempo, Free radical is used as a useful Spin Trap for in Vitro Experiments. Reaction Between Tempone And Biologically Significant Reactive Oxygen Species can be monitored By Esr Spectroscopy. It is also used In the synthesis And Reactions Of Stable Nitroxyl Radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Oxo-Tempo, Free radical is used as a useful Spin Trap for in Vitro Experiments. Reaction Between Tempone And Biologically Significant Reactive Oxygen Species can be monitored By Esr Spectroscopy. It is also used In the synthesis And Reactions Of Stable Nitroxyl Radicals.

Solubility
Soluble in water (partly), acetone, DMSO, 100% ethanol, and diethylether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text