9-Chloroanthracene, is used as a chlorinated anthracene compound used for biochemical research. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Chloroanthracene, is used as a chlorinated anthracene compound used for biochemical research. It is also used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
General References:
- VD Kiselev.; EA Kashaeva.; AI Konovalov. Pressure effect on the rate and equilibrium constants of the Diels-Alder reaction 9-chloroanthracene with tetracyanoethylene. Tetrahedron. 199955(4) , 1153-1162.
- DC Nonhebel. 9-Chloroanthracene. Organic Syntheses. 19635(1) , 115-145.