Thermo Scientific Chemicals

(Bromomethyl)cyclopropane, 97%, Thermo Scientific Chemicals

Catalog number: L01122.03
1 g, Each
Thermo Scientific Chemicals

(Bromomethyl)cyclopropane, 97%, Thermo Scientific Chemicals

Catalog number: L01122.03
1 g, Each
Quantity
Catalog number: L01122.03
also known as L01122-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7051-34-5
IUPAC Name
(bromomethyl)cyclopropane
Molecular Formula
C4H7Br
InChI Key
AEILLAXRDHDKDY-UHFFFAOYSA-N
SMILES
BrCC1CC1
Appearance (Color)
Clear, colourless to pale yellow
Form
liquid
Assay (GC)
≥ 96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4745 - 1.4795 @20?C

Description

(Bromomethyl)cyclopropane is used in the synthesis of 1,4-dienes by iron-catalyzed cross-coupling with alkenyl Grignard reagents. It has been used as a synthetic building block for the introduction of the cyclopropylmethyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Bromomethyl)cyclopropane is used in the synthesis of 1,4-dienes by iron-catalyzed cross-coupling with alkenyl Grignard reagents. It has been used as a synthetic building block for the introduction of the cyclopropylmethyl group.

Solubility
Not miscible in water.

Notes
Protect from heat. Store away from oxidizing agents. Incompatible with strong bases, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text