(Cyclopropylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals
(Cyclopropylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Cyclopropylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01145.06
also known as L01145-06
Price (USD)/ Each
110.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
110.00
Add to cart
(Cyclopropylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals
Catalog numberL01145.06
Price (USD)/ Each
110.00
-
Add to cart
Chemical Identifiers
CAS14799-82-7
IUPAC Name(cyclopropylmethyl)triphenylphosphanium bromide
Molecular FormulaC22H22BrP
InChI KeyWFQSHRSBITUSIB-UHFFFAOYSA-M
SMILES[Br-].C(C1CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water Content (Karl Fischer Titration)≤1%
Assay (Titration ex Bromide)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)177.0-183.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only