Thermo Scientific Chemicals

N-Benzyl-tert-butylamine, 96%, Thermo Scientific Chemicals

Catalog number: L01146.22
100 g, Each
Thermo Scientific Chemicals

N-Benzyl-tert-butylamine, 96%, Thermo Scientific Chemicals

Catalog number: L01146.22
100 g, Each
Quantity
Catalog number: L01146.22
also known as L01146-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3378-72-1
IUPAC Name
benzyl(tert-butyl)azanium
Molecular Formula
C11H18N
InChI Key
DLSOILHAKCBARI-UHFFFAOYSA-O
SMILES
CC(C)(C)[NH2+]CC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥95.0%
Form
Liquid or viscous liquid
Identification (FTIR)
Conforms
Refractive Index
1.4940-1.5000 @ 20?C

Description

N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text