2,4,6-Tribromotoluene, 98+%, Thermo Scientific Chemicals
2,4,6-Tribromotoluene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Tribromotoluene, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L01149.18
also known as L01149-18
Price (USD)/ Each
234.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
234.00
Add to cart
2,4,6-Tribromotoluene, 98+%, Thermo Scientific Chemicals
Catalog numberL01149.18
Price (USD)/ Each
234.00
-
Add to cart
Chemical Identifiers
CAS6320-40-7
IUPAC Name1,3,5-tribromo-2-methylbenzene
Molecular FormulaC7H5Br3
InChI KeyBFRIZWKDNUHPHL-UHFFFAOYSA-N
SMILESCC1=C(Br)C=C(Br)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder or lumps or needles
Melting Point (clear melt)65.0-72.0?C
Appearance (Color)White to pale cream to cream to brown
2,4,6-Tribromotoluene, is used as a chemical and organic intermediate. It can be used to produce 2,4,6-tribromo-benzyl bromide at temperature of 200 - 220°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Tribromotoluene, is used as a chemical and organic intermediate. It can be used to produce 2,4,6-tribromo-benzyl bromide at temperature of 200 - 220°C.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. L Lai.; G Huang.; X Wang.; J Weng. Solvothermal syntheses of hollow carbon microspheres modified with-NH 2 and-OH groups in one-step process. Carbon. 201048(11) , 3145-3156.
  2. H Nishide.; M Nambo.; M Miyasaka. Hyperbranched poly (phenylenevinylene) bearing pendant phenoxys for a high-spin alignment. Journal of Materials Chemistry. 20021(12) , 3578-3584.