2,4-Nonanedione, 98+%, Thermo Scientific Chemicals
2,4-Nonanedione, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Nonanedione, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L01151.14
also known as L01151-14
Price (USD)/ Each
370.65
Online exclusive
412.00 
Save 41.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
370.65
Online exclusive
412.00 
Save 41.35 (10%)
Add to cart
2,4-Nonanedione, 98+%, Thermo Scientific Chemicals
Catalog numberL01151.14
Price (USD)/ Each
370.65
Online exclusive
412.00 
Save 41.35 (10%)
-
Add to cart
Chemical Identifiers
CAS6175-23-1
IUPAC Namenonane-2,4-dione
Molecular FormulaC9H16O2
InChI KeyKFBXUKHERGLHLG-UHFFFAOYSA-N
SMILESCCCCCC(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.4585-1.4625 @ 20?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only