Thermo Scientific Chemicals

3-Iodoaniline, 98%, Thermo Scientific Chemicals

Catalog number: L01159.09
10 g, Each
Thermo Scientific Chemicals

3-Iodoaniline, 98%, Thermo Scientific Chemicals

Catalog number: L01159.09
10 g, Each
Quantity
Catalog number: L01159.09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
626-01-7
IUPAC Name
3-iodoaniline
Molecular Formula
C6H6IN
InChI Key
FFCSRWGYGMRBGD-UHFFFAOYSA-N
SMILES
NC1=CC=CC(I)=C1
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to yellow to red to brown
Form
Fused solid or liquid as melt
Refractive Index
1.6810-1.6870 as melt

Description

3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store at 4°C. Store protected from light. Incompatible with Acid chlorides, acid anhydrides, acids, chloroformates, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text