1-Dodecanesulfonyl chloride, 97%, Thermo Scientific Chemicals
1-Dodecanesulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Dodecanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L01224.14
also known as L01224-14
Price (USD)/ Each
290.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
290.00
Add to cart
1-Dodecanesulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog numberL01224.14
Price (USD)/ Each
290.00
-
Add to cart
Chemical Identifiers
CAS10147-40-7
IUPAC Namedodecane-1-sulfonyl chloride
Molecular FormulaC12H25ClO2S
InChI KeyPBULHKIPTBIZHO-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only