Thermo Scientific Chemicals

2-Amino-1,3,4-thiadiazole, 98+%, Thermo Scientific Chemicals

Catalog number: L01244.14
25 g, Each
Thermo Scientific Chemicals

2-Amino-1,3,4-thiadiazole, 98+%, Thermo Scientific Chemicals

Catalog number: L01244.14
25 g, Each
Quantity
Catalog number: L01244.14
also known as L01244-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4005-51-0
IUPAC Name
1,3,4-thiadiazol-2-amine
Molecular Formula
C2H3N3S
InChI Key
QUKGLNCXGVWCJX-UHFFFAOYSA-N
SMILES
NC1=NN=CS1
Appearance (Color)
White to grey or cream
Form
Crystalline powder
Assay (HPLC)
>98.0%

Description

2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.

Solubility
It is soluble in water, hot methanol- very faint turbidity.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text