Thermo Scientific Chemicals

3-(3-Hydroxyphenyl)propionic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L01279.14
25 g, Each
Thermo Scientific Chemicals

3-(3-Hydroxyphenyl)propionic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L01279.14
25 g, Each
Quantity
Catalog number: L01279.14
also known as L01279-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
621-54-5
IUPAC Name
3-(3-hydroxyphenyl)propanoic acid
Molecular Formula
C9H10O3
InChI Key
QVWAEZJXDYOKEH-UHFFFAOYSA-N
SMILES
OC(=O)CCC1=CC=CC(O)=C1
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥98.0%
Melting Point (clear melt)
110.0-116.0?C
Appearance (Color)
Cream to brown
Identification (FTIR)
Conforms

Description

3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.

Solubility
Soluble in chloroform and methanol.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text