2-Methoxybenzonitrile, 98%, Thermo Scientific Chemicals
2-Methoxybenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxybenzonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number L01293.14
also known as L01293-14
Price (USD)/ Each
116.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
116.00
Add to cart
2-Methoxybenzonitrile, 98%, Thermo Scientific Chemicals
Catalog numberL01293.14
Price (USD)/ Each
116.00
-
Add to cart
Chemical Identifiers
CAS6609-56-9
IUPAC Name2-methoxybenzonitrile
Molecular FormulaC8H7NO
InChI KeyFSTPMFASNVISBU-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5440-1.5480 @ 20?C
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only