3-(3-Methoxyphenyl)propionic acid is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(3-Methoxyphenyl)propionic acid is used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- Wayne K. Anderson.; Arvela R. Heider.; Natarajan Raju.; Jeffery A. Yucht. Vinylogous carbinolamine tumor inhibitors. 23. Synthesis and antileukemic activity of bis[[(carbamoyl)oxy]methyl]-substituted pyrrolo[2,1-a]isoquinolines, pyrrolo[1,2-a]quinolines, pyrrolo[2,1-a]isobenzazepines, and pyrrolo[1,2-a]benzazepines. J. Med. Chem. 1988, 31, (11), 2097-2102.
- Owen B. Wallace.; David W. Smith.; Milind S. Deshpande.; Craig Polson.; Kevin M. Felsenstein. Inhibitors of Aβ production: solid-phase synthesis and SAR of α-hydroxycarbonyl derivatives. Bioorganic & Medicinal Chemistry Letters. 2003, 13, (6), 1203-1206.