Thermo Scientific Chemicals

4-Fluorophenylurea, 96%, Thermo Scientific Chemicals

Catalog number: L01314.06
5 g, Each
Thermo Scientific Chemicals

4-Fluorophenylurea, 96%, Thermo Scientific Chemicals

Catalog number: L01314.06
5 g, Each
Quantity
Catalog number: L01314.06
also known as L01314-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
659-30-3
IUPAC Name
(4-fluorophenyl)urea
Molecular Formula
C7H7FN2O
InChI Key
IQZBVVPYTDHTIP-UHFFFAOYSA-N
SMILES
NC(=O)NC1=CC=C(F)C=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
177.0-186.0?C
Appearance (Color)
White to pale cream
Assay (HPLC)
≥95.0%

Description

4-Fluorophenylurea is widely used as a filter for drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylurea is widely used as a filter for drug.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text