4,4'-Dimethoxydiphenylamine is a diphenylamine derivative used as a chemical additive for cured rubber. It is used in the synthesis of amine electron donating groups in organic donor-bridge-acceptor dyads in Dye Sensitized Solar Cells (DSSC). 4,4?-Dimethoxydiphenylamine is an electron donor and used in the synthesis of organic nonlinear optical (NLO) chromophores which exhibit high thermal and chemical stability, high degree of non-linearity and transparency. They are used in high performance photonic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Dimethoxydiphenylamine is a diphenylamine derivative used as a chemical additive for cured rubber. It is used in the synthesis of amine electron donating groups in organic donor-bridge-acceptor dyads in Dye Sensitized Solar Cells (DSSC). 4,4′-Dimethoxydiphenylamine is an electron donor and used in the synthesis of organic nonlinear optical (NLO) chromophores which exhibit high thermal and chemical stability, high degree of non-linearity and transparency. They are used in high performance photonic devices.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Air Sensitive. Store under dry inert gas. Incompatible with isocyanates, halogenated organics, peroxides, phenols (acidic), epoxides, anhydrides, and acid halides.
RUO – Research Use Only
Joshua A. Davies; Arumugasamy Elangovan; Philip A. Sullivan; Benjamin C. Olbricht; Denise H. Bale; Todd R. Ewy; Christine M. Isborn; Bruce E. Eichinger; Bruce H. Robinson; Philip J. Reid; Xiaosong Li and Larry R. Dalton. Rational Enhancement of Second-Order Nonlinearity: Bis-(4-methoxyphenyl)hetero-aryl-amino Donor-Based Chromophores: Design, Synthesis, and Electrooptic Activity.J. Am. Chem. Soc.2008, 130 (32), 10565-10575.