Thermo Scientific Chemicals

2-(2-Pyridyl)benzimidazole, 98+%, Thermo Scientific Chemicals

Catalog number: L01339.06
5 g, Each
Thermo Scientific Chemicals

2-(2-Pyridyl)benzimidazole, 98+%, Thermo Scientific Chemicals

Catalog number: L01339.06
5 g, Each
Quantity
Catalog number: L01339.06
also known as L01339-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1137-68-4
IUPAC Name
2-(pyridin-2-yl)-1H-1,3-benzodiazole
Molecular Formula
C12H9N3
InChI Key
YNFBMDWHEHETJW-UHFFFAOYSA-N
SMILES
N1C2=CC=CC=C2N=C1C1=CC=CC=N1
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Appearance (Color)
White to cream to yellow to brown
Form
Crystals or powder or crystalline powder

Description

2-(2-Pyridyl)benzimidazole is used as a chelating fluorescent ligand in the synthesis of cobalt(II) complex. It was also used in the preparation of (H(2)L(1))(2)[Ru(III)Cl(4)(CH(3)CN)(2)](2)[Ru(IV)Cl(4)(CH(3)CN)(2)]·2Cl·6H(2)O complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Pyridyl)benzimidazole is used as a chelating fluorescent ligand in the synthesis of cobalt(II) complex. It was also used in the preparation of (H(2)L(1))(2)[Ru(III)Cl(4)(CH(3)CN)(2)](2)[Ru(IV)Cl(4)(CH(3)CN)(2)]·2Cl·6H(2)O complex.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text