2-Mercaptoimidazole, 98+%, Thermo Scientific Chemicals
2-Mercaptoimidazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Mercaptoimidazole, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number L01346.14
also known as L01346-14
Price (USD)/ Each
529.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
529.00
Add to cart
2-Mercaptoimidazole, 98+%, Thermo Scientific Chemicals
Catalog numberL01346.14
Price (USD)/ Each
529.00
-
Add to cart
Chemical Identifiers
CAS872-35-5
IUPAC Name1H-imidazole-2-thiol
Molecular FormulaC3H4N2S
InChI KeyOXFSTTJBVAAALW-UHFFFAOYSA-N
SMILESSC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Assay (HPLC)≥98.0%
Melting Point (clear melt)221-228?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only