1-Naphthalenemethanol, 98+%, Thermo Scientific Chemicals
1-Naphthalenemethanol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Naphthalenemethanol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L01347.09
also known as L01347-09
Price (USD)/ Each
52.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
52.00
Add to cart
1-Naphthalenemethanol, 98+%, Thermo Scientific Chemicals
Catalog numberL01347.09
Price (USD)/ Each
52.00
-
Add to cart
Chemical Identifiers
CAS4780-79-4
IUPAC Name(naphthalen-1-yl)methanol
Molecular FormulaC11H10O
InChI KeyPBLNHHSDYFYZNC-UHFFFAOYSA-N
SMILESOCC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystalline powder or powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)57-64?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only