Cinnamyl bromide, is used in the synthesis of naftifine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cinnamyl bromide, is used in the synthesis of naftifine.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
C.B. Kanner.; U.K. Pandit. Functionalized enamines—XXXII: Reaction of β-amino-α, β-unsaturated esters and amides with benzyl and cinnamyl bromides. Tetrahedron. 1982, 38, (24), 3597-3604.
Methvin B.Isaac.; Tak-Hang Chan. Indium mediated coupling of aldehydes with allyl bromides in aqueous media. the issue of regio- and diastereo-selectivity. Tetrahedron Letters. 1995, 36, (49), 8957-8960.