Thermo Scientific Chemicals

1,4-Benzodioxan-6-amine, 99%, Thermo Scientific Chemicals

Catalog number: L01421.14
25 g, Each
Thermo Scientific Chemicals

1,4-Benzodioxan-6-amine, 99%, Thermo Scientific Chemicals

Catalog number: L01421.14
25 g, Each
Quantity
Catalog number: L01421.14
also known as L01421-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22013-33-8
IUPAC Name
2,3-dihydro-1,4-benzodioxin-6-amine
Molecular Formula
C8H9NO2
InChI Key
BZKOZYWGZKRTIB-UHFFFAOYSA-N
SMILES
NC1=CC=C2OCCOC2=C1
Assay (GC)
≥98.5%
Appearance (Color)
Pale yellow to yellow to dark red to dark brown to brown to black
Form
Crystals or powder or crystalline powder or fused solid or clear liquid or viscous liquid as melt
Refractive Index
1.5985-1.6015 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text