Thermo Scientific Chemicals

5-Nitrothiophene-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: L01493.06
5 g, Each
Thermo Scientific Chemicals

5-Nitrothiophene-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: L01493.06
5 g, Each
Quantity
Catalog number: L01493.06
also known as L01493-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4521-33-9
IUPAC Name
5-nitrothiophene-2-carbaldehyde
Molecular Formula
C5H3NO3S
InChI Key
CHTSWZNXEKOLPM-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(S1)C=O
Form
Crystals or powder or crystalline powder or lumps or fused solid or needles
Free acid (titration)
≤1.5%
Appearance (Color)
Cream to yellow to green to brown to gray
Assay (GC)
≥97.5%

Description

5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text