Methyl vinyl sulfone, 95%, stab. with 200ppm 4-tert-butylphenol, Thermo Scientific Chemicals
Methyl vinyl sulfone, 95%, stab. with 200ppm 4-tert-butylphenol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl vinyl sulfone, 95%, stab. with 200ppm 4-tert-butylphenol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L01519.06
also known as L01519-06
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
Methyl vinyl sulfone, 95%, stab. with 200ppm 4-tert-butylphenol, Thermo Scientific Chemicals
Catalog numberL01519.06
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS3680-02-2
IUPAC Namemethanesulfonylethene
Molecular FormulaC3H6O2S
InChI KeyWUIJTQZXUURFQU-UHFFFAOYSA-N
SMILESCS(=O)(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Solution Test10% v/v in methanol: Clear
Appearance (Color)Clear, colorless to pale yellow
Assay (GC)>94.0%
Refractive Index1.4610-1.4650 @ 20?C
Solution Test10% v/v in diethyl ether: Clear
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only