Thermo Scientific Chemicals

3,4,5-Trimethoxyphenol, 97%, Thermo Scientific Chemicals

Catalog number: L01527.14
25 g, Each
Thermo Scientific Chemicals

3,4,5-Trimethoxyphenol, 97%, Thermo Scientific Chemicals

Catalog number: L01527.14
25 g, Each
Quantity
Catalog number: L01527.14
also known as L01527-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
642-71-7
IUPAC Name
3,4,5-trimethoxyphenol
Molecular Formula
C9H12O4
InChI Key
VTCDZPUMZAZMSB-UHFFFAOYSA-N
SMILES
COC1=CC(O)=CC(OC)=C1OC
Appearance (Color)
White to cream to yellow to brown
Form
Powder
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
142-149?C

Description

3,4,5-Trimethoxyphenol is used as Flavonoid drug intermediates. It is used for pharmaceutical, cosmetics, agrochemicals and food industry research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxyphenol is used as Flavonoid drug intermediates. It is used for pharmaceutical, cosmetics, agrochemicals and food industry research.

Solubility
Soluble in chloroform.

Notes
Light Sensitive. Store in the dark. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text