Thermo Scientific Chemicals

Trifluoroacetaldehyde ethyl hemiacetal, tech. 80%, Thermo Scientific Chemicals

Catalog number: L01554.06
5 g, Each
Thermo Scientific Chemicals

Trifluoroacetaldehyde ethyl hemiacetal, tech. 80%, Thermo Scientific Chemicals

Catalog number: L01554.06
5 g, Each
Quantity
Catalog number: L01554.06
also known as L01554-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
433-27-2
IUPAC Name
1-ethoxy-2,2,2-trifluoroethan-1-ol
Molecular Formula
C4H7F3O2
InChI Key
KLXJPQNHFFMLIG-UHFFFAOYNA-N
SMILES
CCOC(O)C(F)(F)F
Appearance (Color)
Clear, colourless
Form
liquid
Assay (GC)
> 78.0%
Identification (FTIR)
Conforms to reference
Refractive Index
1.3370 - 1.3470 @20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text