Trifluoroacetaldehyde ethyl hemiacetal, tech. 80%, Thermo Scientific Chemicals
Trifluoroacetaldehyde ethyl hemiacetal, tech. 80%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetaldehyde ethyl hemiacetal, tech. 80%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L01554.14
also known as L01554-14
Price (USD)/ Each
257.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
257.00
Add to cart
Trifluoroacetaldehyde ethyl hemiacetal, tech. 80%, Thermo Scientific Chemicals
Catalog numberL01554.14
Price (USD)/ Each
257.00
-
Add to cart
Chemical Identifiers
CAS433-27-2
IUPAC Name1-ethoxy-2,2,2-trifluoroethan-1-ol
Molecular FormulaC4H7F3O2
InChI KeyKLXJPQNHFFMLIG-UHFFFAOYNA-N
SMILESCCOC(O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Appearance (Color)Clear, colourless
Assay (GC)> 78.0%
Identification (FTIR)Conforms to reference
Refractive Index1.3370 - 1.3470 @20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only