Valeric acid hydrazide, 95%, Thermo Scientific Chemicals
Valeric acid hydrazide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Valeric acid hydrazide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L01555.14
also known as L01555-14
Price (USD)/ Each
83.90
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
83.90
Add to cart
Valeric acid hydrazide, 95%, Thermo Scientific Chemicals
Catalog numberL01555.14
Price (USD)/ Each
83.90
-
Add to cart
Chemical Identifiers
CAS38291-82-6
IUPAC Namepentanehydrazide
Molecular FormulaC5H12N2O
InChI KeyPJBQYCIDGYKEMN-UHFFFAOYSA-N
SMILESCCCCC(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)50.5-62.5?C
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only