D-(-)-2-Phenylglycine ethyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
D-(-)-2-Phenylglycine ethyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-(-)-2-Phenylglycine ethyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L01564.09
also known as L01564-09
Price (USD)/ Each
97.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
97.00
Add to cart
D-(-)-2-Phenylglycine ethyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
Catalog numberL01564.09
Price (USD)/ Each
97.00
-
Add to cart
Chemical Identifiers
CAS17609-48-2
IUPAC Namehydrogen ethyl 2-(2-aminophenyl)acetate chloride
Molecular FormulaC10H14ClNO2
InChI KeyQITKMRXHUSYVSD-UHFFFAOYSA-N
SMILES[H+].[Cl-].CCOC(=O)CC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
FormPowder or granules
Optical Rotation-93.6? ± 0.5? (c=1 in water)
D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only