Thermo Scientific Chemicals

D-(-)-2-Phenylglycine ethyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: L01564.18
50 g, Each
Thermo Scientific Chemicals

D-(-)-2-Phenylglycine ethyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: L01564.18
50 g, Each
Quantity
Catalog number: L01564.18
also known as L01564-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17609-48-2
IUPAC Name
hydrogen ethyl 2-(2-aminophenyl)acetate chloride
Molecular Formula
C10H14ClNO2
InChI Key
QITKMRXHUSYVSD-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CCOC(=O)CC1=CC=CC=C1N
Form
Powder or granules
Optical Rotation
-93.6? ± 0.5? (c=1 in water)
Appearance (Color)
White
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%

Description

D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text