3,4,5-Trimethoxyaniline, 98+%, Thermo Scientific Chemicals
3,4,5-Trimethoxyaniline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4,5-Trimethoxyaniline, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L01619.14
also known as L01619-14
Price (USD)/ Each
125.65
Online exclusive
139.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
125.65
Online exclusive
139.00 
Save 13.35 (10%)
Add to cart
3,4,5-Trimethoxyaniline, 98+%, Thermo Scientific Chemicals
Catalog numberL01619.14
Price (USD)/ Each
125.65
Online exclusive
139.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS24313-88-0
IUPAC Name3,4,5-trimethoxyaniline
Molecular FormulaC9H13NO3
InChI KeyXEFRNCLPPFDWAC-UHFFFAOYSA-N
SMILESCOC1=CC(N)=CC(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream to pale brown or pale pink or pale yellow
Melting Point (clear melt)110.0-116.0?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only