Thermo Scientific Chemicals

2,5-Dimethylbenzyl alcohol, 97+%, Thermo Scientific Chemicals

Catalog number: L01624.03
1 g, Each
Thermo Scientific Chemicals

2,5-Dimethylbenzyl alcohol, 97+%, Thermo Scientific Chemicals

Catalog number: L01624.03
1 g, Each
Quantity
Catalog number: L01624.03
also known as L01624-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53957-33-8
IUPAC Name
(2,5-dimethylphenyl)methanol
Molecular Formula
C9H12O
InChI Key
LEBQTCCCNMTXSF-UHFFFAOYSA-N
SMILES
CC1=CC=C(C)C(CO)=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥97.0%
Form
Liquid
Refractive Index
1.5335-1.5385 @ 20?C

Description

2,5-Dimethylbenzyl alcohol, is used as a self-indicating standard for the determination of organ lithium reagents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylbenzyl alcohol, is used as a self-indicating standard for the determination of organ lithium reagents

Solubility
Difficult in mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text