1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.
Solubility
Not miscible in water.
Notes
Store away from oxidizing agents, reducing agents and water. Hygroscopic.
RUO – Research Use Only
General References:
- Mary Frances Richardson; Robert E. Sievers. Volatile rare earth chelates of 1,1,1,5,5,5-hexafluoro-2,4-pentanedione and 1,1,1,2,2,3,3,7,7,7-decafluoro-4,6-heptanedione.Inorg. Chem.1971, 10 (3), 498-504.
- Richard Steger; Rich Masel. Chemical vapor etching of copper using oxygen and 1,1,1,5,5,5-hexafluoro-2,4-pentanedione.Thin Solid Films.1999, 342 221-229.